Learn More
Kurin Jet (Blood Culture Collection Device)
SDP
Jet is a blood culture (BC) collection device which significantly lowers BC contamination rates. Jet automatically sidelines the initial 0.15 mL of blood, which may contain contaminants from the patient’s own skin, ensuring that the best possible blood specimen reaches the collection bottles.
Supplier:  Kurin TJ221
 View more versions of this product
Catalog No. 23-232-021
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Description
Description
  • Lowers blood culture contamination rate
  • Helps achieve <1% contamination rate
  • Minimal blood volume waste
  • Closed System - All-In-One Device
  • Fits BD, bioMerieux & ThermoFisher Blood Culture Bottles
Specifications
Specifications
Jet with21 G Push Button Safety Needle for ThermoFisher Blood Culture Bottles (case)
Blood Culture Collection
200 per Cs.
PushButton Safety Needle, Kurin Jet, Vacutainer Shield to fit Thermo BC Bottles
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.