Learn More
Gold Biotechnology Inc JM109 Chemically Competent E. coli Cells 10x50 μl
SDP
Supplier:  Gold Biotechnology Inc CC11410X50
Encompass_Preferred
GoldBio’s JM109 Chemically Competent E. coli cells are suitable for high efficiency transformation in a wide variety of routine applications such as plasmid isolation, library construction, cloning, and subcloning. JM109 is a K strain bacterium that carries multiple features including the lacIqZΔM15 marker, which provides α-complementation of the β-galactosidase gene with blue/white screening protocol as well as the hsdR17 genotype which prevents endogenous endonucleases from cleaving DNA.
Catalog No. NC2589336
missing translation for 'QuantityLeft'
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.