Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
JNJ 10397049, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
Selective OX2 receptor antagonist (pIC50 = 7.4 for chimeric OX2 receptors; pKB values are 5.9 and 8.5 for OX1 and OX2 receptors respectively). Shows no significant activity in a panel of over 50 other neurotransmitters and neuropeptide receptors. Achieves high level of OX2 receptor occupancy in the rat brain; exhibits sleep-promoting effects in rats.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 484.18 |
Percent Purity | >99% |
Chemical Name or Material | JNJ 10397049 |
Safety and Handling
Recommended Storage | Store at +4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction