Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JP 1302 dihydrochloride, Tocris Bioscience™

Catalog No. p-4784059 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective α2C antagonist

α2C-adrenoceptor antagonist that displays ∽ 50-fold selectivity over other α2-adrenoceptor subtypes (Ki values are 28, 1470, 1700 and 3150 nM for human α2C, α2B, α2D and α2A subtypes respectively). Potently antagonizes adrenalin-stimulated 35GTPγS binding in vitro (KB = 16 nM) and produces antidepressant and antipsychotic-like effects in vivo.

Chemical Identifiers

CAS 1259314-65-2
Molecular Formula C24H26Cl2N4
Molecular Weight (g/mol) 441.4
InChI Key VVZOADYFJAJZGL-UHFFFAOYSA-N
Synonym jp 1302 dihydrochloride, n-4-4-methyl-1-piperazinyl phenyl-9-acridinamine dihydrochloride, n-4-4-methyl-1-piperazinyl phenyl-9-acridinaminedihydrochloride, n-4-4-methylpiperazin-1-yl phenyl acridin-9-amine dihydrochloride
PubChem CID 49855035
IUPAC Name N-[4-(4-methylpiperazin-1-yl)phenyl]acridin-9-amine;dihydrochloride
SMILES CN1CCN(CC1)C2=CC=C(C=C2)NC3=C4C=CC=CC4=NC5=CC=CC=C53.Cl.Cl

Specifications

Quantity 10 mg
Formula Weight 441.4
Percent Purity >99%
Chemical Name or Material JP 1302 dihydrochloride
Recommended Storage Store at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.