Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Justrite™ Base Cover for 90-Minute EN Cabinets

Catalog No. p-6762006 Shop All Justrite Safety Group Products
Click to view available options
For Use With (Equipment):

Keeps debris out from underneath cabinets by adding an optional, removeable base cover

  • Made from textured, durable powder-coated steel
  • Conceals integrated transport base on cabinet
  • Prevents dust and debris from collecting underneath cabinet
  • Creates seamless flush-to-floor attractive presentation
  • Yellow

Specifications

Material

Steel

For Use With (Equipment)

30 gal. EN cabinet

Surface Coating

Powder-coated

Description

Prevents collection of dust, debris; Textured, durable powder-coated steel

Color

Yellow

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.