Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JZL 184, Tocris Bioscience™

Catalog No. p-4816565 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent MAGL inhibitor

Potent and selective MAGL inhibitor. Blocks hydrolysis of the endocannabinoid 2-arachidonyl glycerol (2-AG) in vivo in the mouse brain (IC50 = 8 nM). Potentiates depolarization-induced suppression of excitability in cerebellar Purkinje neurons. Exhibits >300-fold selectivity for MAGL over FAAH in vitro. Attenuates nociception in neuropathic and inflammatory pain models. Also reduces free fatty acid levels in primary tumors.

Chemical Identifiers

CAS 1101854-58-3
Molecular Formula C27H24N2O9
Molecular Weight (g/mol) 520.494
InChI Key SEGYOKHGGFKMCX-UHFFFAOYSA-N
Synonym unii-7mz1i2j68a, 4-nitrophenyl 4-bis 2h-1,3-benzodioxol-5-yl hydroxy methyl piperidine-1-carboxylate, 4-nitrophenyl 4-dibenzo d 1,3 dioxol-5-yl hydroxy methyl piperidine-1-carboxylate, 1-piperidinecarboxylic acid, 4-bis 1,3-benzodioxol-5-yl hydroxymethyl-, 4-nitrophenyl ester, d0g0mz, 4-bis 1,3-benzodioxol-5-yl hydroxymethyl-1-piperidinecarboxylic acid 4-nitrophenyl ester, 4-nitrophenyl 4-bis benzo d 1,3 dioxol-5-yl hydroxy methyl piperidine-1-carboxylate, 4-nitrophenyl 4-dibenzo d 1,3 dioxol-5-yl-hydroxy methyl piperidine-1-carboxylate, 4-nitrophenyl 4-bis 1,3-benzodioxol-5-yl hydroxy methyl-1-piperidinecarboxylate
PubChem CID 25021165
IUPAC Name (4-nitrophenyl) 4-[bis(1,3-benzodioxol-5-yl)-hydroxymethyl]piperidine-1-carboxylate
SMILES C1CN(CCC1C(C2=CC3=C(C=C2)OCO3)(C4=CC5=C(C=C4)OCO5)O)C(=O)OC6=CC=C(C=C6)[N+](=O)[O-]

Specifications

Quantity 10 mg
Formula Weight 520.49
Percent Purity >98%
Chemical Name or Material JZL 184
Recommended Storage Store at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.