Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JZP 361, >99%, Tocris Bioscience™

Catalog No. p-7121260 Shop All R&D Systems Products
Click to view available options
Quantity:
10mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective reversible MAGL inhibitor

Potent and selective reversible MAGL inhibitor (IC50 = 46 nM). Exhibits 35-fold and 150-fold higher selectivity over human FAAH and ABHD6, respectively (IC50 values are 1.79 and 7.24 μM, respectively). Displays affinity for H1 receptors (pA2 = 6.81). Exhibits no activity at cannabinoid receptors at 10 μM.

Specifications

Chemical Name or Material [4-(8-Chloro-5,6-dihydro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-ylidene)-1-piperidinyl](1H-1,2,4-
CAS 1680193-80-9
Purity Grade Notes >99%
Molecular Formula C22H20ClN5O
Linear Formula C22H20ClN5O
Quantity 10mg
Synonym 4-8-chloro-5,6-dihydro-11h-benzo 5,6 cyclohepta 1,2-b pyridin-11-ylidene-1-piperidinyl 1h-1,2,4-triazol-1-yl methanone
Solubility Information Soluble to 100mM in DMSO and to 20mM in ethanol
InChI Key GAVZCGTYRWKKDV-UHFFFAOYSA-N
SMILES C1CC2=C(C=CC(=C2)Cl)C(=C3CCN(CC3)C(=O)N4C=NC=N4)C5=C1C=CC=N5
IUPAC Name [4-(8-chloro-5,6-dihydrobenzo[1,2]cyclohepta[2,4-b]pyridin-11-ylidene)piperidin-1-yl]-(1,2,4-triazol-1-yl)methanone
Molecular Weight (g/mol) 405.886
PubChem CID 102596130
Formula Weight 405.88
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.