Learn More
Neogen Diagnostics K-BLUE AQUEOUS (TMB) - 200 ML
Supplier:  Neogen Diagnostics 331175
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
TMB substrate for HRP microwell applications. 100% solvent free. Ready-to-use.
Catalog No. 50-134-9569
missing translation for 'QuantityLeft'