Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kallikrein 5 Mouse anti-Human, Clone: KLK5/3841, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP307858100UG
Description
Kallikrein 5 Monoclonal specifically detects Kallikrein 5 in Human samples. It is validated for Immunohistochemistry-Paraffin, Protein Array.Specifications
Kallikrein 5 | |
Monoclonal | |
0.2 mg/mL | |
Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array | |
EC 3.4.21, EC 3.4.21.4, kallikrein 5, Kallikrein-like protein 2, kallikrein-related peptidase 5, KLKL2, KLK-L2EC 3.4.21.-, SCTEkallikrein-5, Stratum corneum tryptic enzyme | |
Recombinant fragment of human Kallikrein 5 protein (around aa 36-177) (exact sequence is proprietary) | |
100 μg | |
Cancer | |
25818 | |
Store at 4°C. | |
IgG2c κ |
Immunohistochemistry (Paraffin), Peptide Array | |
KLK5/3841 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?