Learn More
Gold Biotechnology Inc Kanamycin Monosulfate, 25389-94-0, MFCD00070253, 100 g
SDP
Supplier:  Gold Biotechnology Inc K120100
Encompass_Preferred
Formula: C18H36N4O11 · H2SO4, MW: 582.58 g/mol, Store desiccated at -20°C, USP Grade, Activity: ~800 µg/mg.
Catalog No. 50-153-2807
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.