Learn More
Gold Biotechnology Inc Kanamycin Monosulfate, USP Grade 5 g
SDP
Supplier:  Gold Biotechnology Inc K1205
Encompass
Kanamycin is an aminoglycoside antibiotic isolated from Streptomyces kanamyceticus. It is effective against gram-positive and gram-negative bacteria as well as Mycoplasma species. Kanamycin is commonly used to select for bacteria that have been transformed with a kanamycin-resistant gene and plant tissues that have the NPT II (APH3) gene incorporated in them. Kanamycin is very soluble in aqueous solution and is typically used in concentrations of 50-100 µg/ml.
Catalog No. NC1835474
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.