Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kappler™ ProVent™ 7000 Isolation Gowns, Tie Closures Neck and Waist
SDP

Catalog No. 19028299 Shop All Kappler, Inc. Products
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Cuff Style:
Elastic
Knit

Breathable, microporous fabric gowns offer general hazard protection for a variety of work environments from maintenance to general healthcare. Choose from elastic wrist or knit cuff options.

Lightweight composite fabric utilizes microporous film technology to provides protection against hazardous particles and light liquid splash and maintain strength wet or dry

  • Wraparound style opens to back
  • Knit cuffs or elastic wrists
  • Two neck ties, two waist ties
  • Nonsterile
  • White

Order Info

  • Front length: small 38.75 in. (98.4 cm), medium 39.75 in. (101 cm), large 40.75 in. (103.5 cm). Back length of all sizes: 3 in. (7.6 cm) longer than front

Compliance

Meets ASTM™ F1670 and ASTM F1671 standards.

Specifications

For Use With (Application) Maintenance, Spray Painting, Healthcare, Clinical
Style Wrap-around
No. of Pockets None
Gender Unisex
Product Type General Purpose Gown
Size X-Large
Material Microporous Coated Fabric
Color White
Closure Type Neck and Waist Ties
Disposable Single-use
Sterility Non-sterile
Cuff Style Elastic
Model PVS112WHXL
Seam Type Serged
Sleeve Style Long Sleeve
Certifications/Compliance ASTM F1670, ASTM F1671
Weight (English) 16 lbs.
Autoclavable Not Autoclavable
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.