Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KB-R7943 mesylate, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent, selective inhibitor of the reverse mode of the Na+/Ca2+ exchanger (IC50 = 0.7 μM). Also inhibits the mitochondrial Ca2+ uniporter (MCU; IC50 = 5.5 μM). Does not affect Na+-dependent transport systems or ionotropic glutamate receptors.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 427.49 |
Percent Purity | >99% |
Chemical Name or Material | KB-R7943 mesylate |
Safety and Handling
Recommended Storage | Store at Room Temperature |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction