Learn More
Sklar Instruments Keyes Cutaneous Punch, Sklar 4 in., 2mm, Straight, Round, Stainless Steel, EA.
Supplier: Sklar Instruments 064142
Keyes Cutaneous Punch, Sklar. 4 in., 2mm, Straight, Round, Stainless Steel. Sklar's Keyes Cutaneous Punch is a dermal diagnostic instrument used to remove skin samples for biopsy. The round sharp end is pushed into the skin and twisted slightly to cut away a small plug of skin. The solid knurled handle provides a gripping surface for the user who controls the depth of the excision. The Keyes punch is often used in gynecological biopsies where a deeper tissue sample is useful for the pathologist. This specific punch has a 2mm diameter, however, is available up to 8mm.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.