Learn More
TARGETMOL CHEMICALS INC KFM19 5MG
Supplier:  TARGETMOL CHEMICALS INC T15655
Encompass_Preferred
Also available in 50 mg 100 mg and bulk. Please contact Fisher for quotes. KFM19 is a potent and selective adenosine receptor (A1-receptor) antagonist (IC50 50 nM) for the study of neurological disorders. purity: 99%
Catalog No. 50-258-1213
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.