Learn More
Kimberly-Clark Professional™ KleenGuard™ V10 Element™ Safety Glasses
When you need an economical way to provide on-the-job eye protection, KleenGuard Element Safety Eyewear is an excellent choice.
Supplier: Kimberly-Clark Professional™ 33072
Description
- Lightweight
- Frameless styling in a metal-free design, complete with screw-less hinges
- Polycarbonate lenses provide 99.9% UVA/ UVB/UVC protection to help prevent eye damage like cataracts, retinal damage, and other conditions that can cause temporary vision loss
- Meet ANSI Z87.1+ standards for high impact in personal eye protection
- Feature a 10 base curve and are TAA compliant
Specifications
Meets ANSI Z87.1+ standards for high impact (33072-01 meets ANSI Z87.1+2010 and 33072-02 meets ANSI Z87.1+2015) | |
Blue | |
Safety Glasses | |
Polycarbonate |