Learn More
Kimberly-Clark Professional™ KleenGuard™ V30 Nemesis™ Small Safety Eyewear
Get everything you love about the original KleenGuard Nemesis Safety Eyewear - stylish good looks and durable, lightweight construction – but in with smaller dimensions.
Supplier: Kimberly-Clark Professional™ 38476
View more versions of this product
Safety Services Available
Overview
Matting Assessments
This item is not returnable.
View return policy
Description
- Durable, lightweight construction
- Offer a sport design that fits smaller, slimmer heads without adjustments
- Features a stronger nose piece and a wider bridge for enhanced comfort
- Polycarbonate lenses that provide 99.9% UVA/ UVB/UVC protection to help prevent eye damage like cataracts, retinal damage, and other conditions that can cause temporary vision loss
- Wraparound frame has a base curve of 8 that extends the range of peripheral protection
- Each pair comes with a neck cord for securing, so they are ready at a moment’s notice
- Manufactured to comply with the ANSI / ISEA Z87.1+ standard for personal eye protection
- Feature a sporty, flexible, lightweight design
- Patented FLEX-DRY designed to be extremely flexible, to channel sweat away from the eyes, and to help prevent slippage down the nose
Specifications
Hardcoat | |
Smoke | |
Black |
Meets ANSI Z87.1+ standards for high impact (38476-00 and 38476-01 meets ANSI Z87.1+2010 and 38476-02 meets ANSI Z87.1+2015) | |
Safety Eyewear | |
Black frame with black tips |