Learn More
Promega Kinase Selectivity Profiling System: CMGC-1 + ADP-Glo™ Assay
Easy-to-use kits for performing kinase selectivity profiling that rely on the ADP-Glo™ Kinase Assay technology. Each system includes kinase and substrate pairs in an easy-to-use 8-tube strip format optimized for fast and simple kinase profiling reactions.
Supplier: Promega V6855
Description
- Selectivity Screening of Library Compounds for Effect on CMGC Kinases
- Kinase and substrate pairs organized in an easy-to-use 8-tube strip format
- Simple reaction assembly and automation-friendly protocol
- Streamilines profiling of hits and leads for potency against families of related kinases
Specifications
8 × 50 reactions | |
384-well Plate | |
Cat.# V6854 Kinase Selectivity Profiling System: CMGC-1, which includes:
|
Components at different temperatures | |
|
|
Kinase Selectivity Profiling System |