Learn More
World Precision Instrument Kite Manual Micromanipulator-KITE-M3-L
Supplier: World Precision Instrument KITEM3L
The KITE is an economical manual micromanipulator that is perfect for student use. It is available in right or left hand versions and is also sold bundled with an optional M3 Tilting Base. Our 5 lb. weight (WPI #5464) is frequently sold with this unit to provide stability.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.