Learn More
Bioworld KLH SEPAROPOREAGAROSE) 4B
Supplier: Bioworld 201810162
KLH-Separopore 4B Affinity matrix with immobilized KLH for the purification of antibodies generated against a peptide-KLH conjugated antigens. It allows removal of antobodies against KLH portion of the antigen; Supplied as a suspension i...
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.