Learn More
KLRG1 Hamster anti-Mouse, BV650, Clone: 2F1, BD Horizon™
Hamster Monoclonal Antibody
Supplier: BD Biosciences 569548
Description
KLRG1 Monoclonal antibody specifically detects KLRG1 in Mouse samples. It is validated for Flow cytometrySpecifications
KLRG1 | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Hamster | |
Affinity Chromatography | |
RUO | |
50928 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. | |
IgG2 κ |
Flow Cytometry | |
2F1 | |
Brilliant Violet 650 | |
Klrg1; Killer cell lectin-like receptor subfamily G member 1; MAFA; 2F1 Ag | |
A-LAK from C57BL/6 mice | |
50 μg | |
Primary | |
Mouse | |
Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.