Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KN-62, Tocris Bioscience™

Catalog No. 12771 Shop All R&D Systems Products
Click to view available options
Quantity:
This item is not returnable. View return policy
This item is not returnable. View return policy

Non-competitive P2X7 antagonist

Selective, cell-permeable inhibitor of CaM kinase II (IC50 = 0.9 μM). Binds directly to the calmodulin binding site of the enzyme. Potent non-competitive antagonist at the P2X7 receptor (IC50 = 15 nM).

Chemical Identifiers

CAS

127191-97-3

Molecular Formula

C38H35N5O6S2

Molecular Weight (g/mol)

721.85

MDL Number

MFCD00083180

InChI Key

RJVLFQBBRSMWHX-UHFFFAOYNA-N

Synonym

s-4-2-n-methylisoquinoline-5-sulfonamido-3-oxo-3-4-phenylpiperazin-1-yl propyl phenyl isoquinoline-5-sulfonate, unii-63hm46xpow, chembl28324, 63hm46xpow, 1-n,o-bis 5-isoquinolinesulfonyl-n-methyl-l-tyrosyl-4-phenylpiperazine, s-5-isoquinolinesulfonic acid 4-2-5-isoquinolinylsulfonyl methylamino-3-oxo-3-4-phenyl-1-piperazinyl propyl phenyl ester, 4-2s-2-5-isoquinolinylsulfonyl methylamino-3-oxo-3-4-phenyl-1-piperazinyl propyl phenyl isoquinolinesulfonic acid ester, 4-2s-2-n-methylisoquinoline-5-sulfonamido-3-oxo-3-4-phenylpiperazin-1-yl propyl phenyl isoquinoline-5-sulfonate, 5-isoquinolinesulfonic acid, 4-2s-2-5-isoquinolinylsulfonyl methylamino-3-oxo-3-4-phenyl-1-piperazinyl propyl phenyl ester

PubChem CID

5312126

IUPAC Name

4-[2-(N-methylisoquinoline-5-sulfonamido)-3-oxo-3-(4-phenylpiperazin-1-yl)propyl]phenyl isoquinoline-5-sulfonate

SMILES

CN(C(CC1=CC=C(OS(=O)(=O)C2=C3C=CN=CC3=CC=C2)C=C1)C(=O)N1CCN(CC1)C1=CC=CC=C1)S(=O)(=O)C1=C2C=CN=CC2=CC=C1

Specifications

Quantity

1 mg

Formula Weight

721.84

Percent Purity

>98%

Chemical Name or Material

KN-62

Recommended Storage

Desiccate at -20°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.