Learn More
AdipoGen Kojic acid
Supplier: AdipoGen BVT0019M010
Chemical. CAS 501-30-4. Formula C6H6O4. MW 142.1. Isolated from Aspergillus sp. Mycotoxin. Chelation agent with polyfunctional skeleton for development of biologically active compounds. Moderate antibacterial agent. Tyrosinase inhibitor. Inhibits catecholase activity of tyrosinase. Anti-melanogenic. Skin whitening agent. Antioxidant. Reactive oxygen species ROS scavenger. D-Amino acid oxidase DAAO inhibitor. Shown to inhibit NF-kB activation. Inducer of macrophage activation. Radioprotectant activity. Antileishmanial agent. Insecticidal. Antifungal chemosensitizer. Used in cosmetics and as food additive.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.