Learn More
DWK Life Sciences Kimble™ Kontes™ Brand Rotary Evaporator Glass Vial Connecting Adapters
Ideal for use with small samples
Supplier:  DWK Life Sciences 7471301314
 View more versions of this product
Catalog No. K747130-1314
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Description
Description
Ideal for use with small samples
Connect standard rotary evaporator traps to vials with variety of GPI thread sizes. Typical receivers include Accuform™ 1 to 10mL vials (06-100C, 06-100E, 06-100F, 06-100G). Any standard vial with GPI threads can be used.
Specifications
Specifications
Standard Taper Joint: 14/20
Glass
13-425
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.