Learn More
Chemglass Life Sciences Krytox Low Vapor Pressure Grease, 2oz Tube
SDP
Supplier:  Chemglass Life Sciences CGLVP01
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Krytox Low Vapor Pressure Grease, 2oz Tube. Highly inert, non-flammable grease for use with standard taper joints and glass stopcocks. Insoluble in almost all solvents except Freon 113. Krytox greases have demonstrated an exceptionally high degree of chemical inertness to many highly reactive chemicals. For example Krytox has shown no evidence of reactivity when exposed to: Boiling Sulfuric Acid, Fluorine Gas, Nitrogen, Tetroxide, Chlorine Trifluoride, Hydrogen Gas, Methanol, Ammonia Gas, Ethyl Alcohol, Hydrochloric Acid, Hydrazine, Diethylenetriamine, Phosphoric Acid, Sulfuric Acid, Chlorine, or Hydrogen Peroxide.LVP Grade:Krytox LVP grease is made with low vapor pressure fluorinated oils and is designed for use in high vacuum applications down to -1.0 x 10-13 Torr at 20 C, and -1.0 x 10-5 Torr at 200 C. The useful temperature range of Krytox LVP is -20 to 260 C. Supplied in convenient 2oz tubes.
Catalog No. CGLVP01
missing translation for 'QuantityLeft'