Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ KS 176
Selective BCRP inhibitor
$242.00 - $1009.00
Specifications
Purity | >98% |
---|---|
Product Type | KS 176 |
Molecular Weight (g/mol) | 405.4 |
Inhibitors | KS 176 |
Description
Selective inhibitor of the breast cancer resistance protein (BCRP) multidrug transporter (IC50 values are 0.59 and 1.39 μM in Pheo A and Hoechst 33342 assays respectively). Displays no inhibitory activity against P-gp or MRP1.Specifications
>98% | |
KS 176 | |
405.4 | |
KS 176 |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title