Learn More
Hach Company Chemical Oxygen Demand (COD) TNTplus Vial Test, HR (20-1,500 mg/L COD), 150 Tests
Supplier:  Hach Company TNT82206
Encompass_Preferred
Hach TNTplus COD vial test reagents are incredibly simple to use and cut down significantly on waste generation compared to other COD test kits. TNT822 is based on Hach Method 8000 for virtually any type of water sample. Method 8000 is USEPA approved for wastewater analysis under 40 CFR 136.3 and based on Standard Methods 5220 D. Hach TNTplus COD vial tests are available in multiple ranges. TNT822 provides a result in the High Range of 20 - 1500 mg/L COD and is often used for Chemical Oxygen Demand measurement in wastewater influent. Hach TNT822 does not require a separate blanking vial for further waste reduction. Make the switch to TNTplus COD reagents today!For determination of Chemical Oxygen Demand (COD) by the Reactor Digestion Method. Simply pipette in sample, digest, and read in your Hach Spectrophotometer. 6x25 tests.
Catalog No. 50-199-8116
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.