Learn More
Hach Company HQ2200 Portable Multi-Meter with pH and Dissolved Oxygen Electrodes, 1 m Cables
Supplier:  Hach Company LEV015.53.22004
Encompass_Preferred
HQ2200 Portable pH/ORP/EC/TDS/DO Meter, 2 channels for waste water plant testing with IntelliCAL PHC201 gel filled pH and LDO101 Dissolved Oxygen electrodes, 1 m Cables. Field accessories included.
Catalog No. 50-206-2958
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.