Learn More
Hach Company Chemical Oxygen Demand (COD) TNTplus Vial Test, UHR (250-15,000 mg/L COD), 150 Tests
Supplier: Hach Company TNT82306
Hach TNTplus COD vial test reagents are incredibly simple to use and cut down significantly on waste generation compared to other COD test kits. TNT823 is based on Hach Method 10212 for virtually any type of water sample. Hach TNTplus COD vial tests are available in multiple ranges. TNT823 provides a result in the Ultra High Range of 250 - 15,000 mg/L COD and is often used for Chemical Oxygen Demand measurement in industrial wastewater influent. Hach TNT823 does not require a separate blanking vial for further waste reduction. Make the switch to TNTplus COD reagents today!For determination of Chemical Oxygen Demand (COD) by the Reactor Digestion Method. Simply pipette in sample, digest, and read in your Hach Spectrophotometer. 6X25 tests.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.