Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KU 0060648, Tocris Bioscience™

Catalog No. p-4816713 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Dual DNA-PK and PI 3-K inhibitor

Dual PI 3-K and DNA-PK inhibitor (IC50 values are <0.1, 0.5, 4 and 19 nM for PI 3-Kδ, PI 3-Kβ, PI 3-Kα and DNA-PK respectively). Inhibits proliferation of MCF7 cells in vitro and delays growth of MCF7 xenografts in mice. Also enhances CRISPR-Cas9-mediated homology-directed repair (HDR) efficiency ∽2 to 4-fold, and decreases nonhomologous end-joining (NHEJ) frequency ∽40%; maximum effect seen at 250 nM.

Chemical Identifiers

CAS 881375-00-4
Molecular Formula C33H34N4O4S
Molecular Weight (g/mol) 582.719
InChI Key AATCBLYHOUOCTO-UHFFFAOYSA-N
Synonym 2-4-ethylpiperazin-1-yl-n-4-2-morpholino-4-oxo-4h-chromen-8-yl dibenzo b,d thiophen-1-yl acetamide, 2-4-ethylpiperazin-1-yl-n-4-2-morpholin-4-yl-4-oxochromen-8-yl dibenzothiophen-1-yl acetamide, d0r0tw, ku hplc, 2-4-ethylpiperazin-1-yl-n-6-2-morpholin-4-yl-4-oxo-4h-chromen-8-yl-8-thiatricyclo 7.4.0.0?,? trideca-1 13 ,2,4,6,9,11-hexaen-3-yl acetamide, 2-4-ethylpiperazin-1-yl-n-6-2-morpholin-4-yl-4-oxochromen-8-yl-8-thiatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaen-3-yl acetamide, 4-ethyl-n-4-2-4-morpholinyl-4-oxo-4h-1-benzopyran-8-yl-1-dibenzothienyl-1-piperazineacetamide, n-4-2-morpholino-4-oxo-4h-1-benzopyran-8-yl dibenzothiophene-1-yl-4-ethylpiperazine-1-acetamide
PubChem CID 11964036
IUPAC Name 2-(4-ethylpiperazin-1-yl)-N-[4-(2-morpholin-4-yl-4-oxochromen-8-yl)dibenzothiophen-1-yl]acetamide
SMILES CCN1CCN(CC1)CC(=O)NC2=C3C4=CC=CC=C4SC3=C(C=C2)C5=CC=CC6=C5OC(=CC6=O)N7CCOCC7

Specifications

Quantity 10 mg
Formula Weight 582.71
Percent Purity >97%
Chemical Name or Material KU 0060648
Recommended Storage Store at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.