Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Kut-Gard™ 07-K320 Seamless Knit Aramid Fiber/Cotton Plated Gloves

Catalog No. p-4891814 Shop All Protective Industrial Products Inc. Products
Click to view available options
Size:
Small
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Cotton-plated aramid fiber construction provides heat resistance, cut protection, and is inherently flame resistant.

  • Aramid fiber construction provides cut protection, heat resistance, and is inherently flame resistant
  • Seamless knit design protects hands without sacrificing comfort or dexterity
  • Material will not melt, ignite, or conduct electricity
  • Ambidextrous pattern reduces replacement costs
  • Color coded hems for easy size identification
  • Can be used alone, or as glove liners

Specifications

Construction Seamless Knit
Coating Material Uncoated
Color Yellow
Cut Resistance Rating ANSI A1
Hand Ambidextrous
Material Cotton, Aramid Fiber
Product Type Seamless Knit Gloves
Size Small
Style Reversible
For Use With (Application) Automotive Assembly, Light Heat, Glass Operations, and Metal Stamping
Glove Cuff Style Knit
Hem Color White
Series 07-K320
Yarn Gauge 7
Length (English) 9 in.
Length (Metric) 22.9 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.