Learn More
Carbosynth LLC. H-Lys-Val-Pro-Arg-Asn-Gln-Asp-Trp-Leu-OH, 1mg, MW: 1155.33, Molecular formula: C52H82N16O14
SDP
Supplier:  Carbosynth LLC. PCP3922PI1MG
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Synonyms: Human GP 100 (25-33), KVPRNQDWL, Application: MHC-class I-Restricted Epitope in hgp100 Heteroclitic MHC Class I Epitope in hgp100, Storage Temperature: -20°C, References: W.W. Overwijk, A. Tsung, K.R. Irvine, M.R. Parkhurst, T.J. Goletz, K. Tsung, M.W. Carroll, C. Liu, B. Moss, S.A. Rosenberg, and N.P. Restifo, J. Exp. Med., 188, 277 (1998)., J.S. Gold, C.R. Ferrone, J.A. G.-Patino, W.G. Hawkins, R. Dyall, M.E. Engelhorn, J.D. Wolchok, J.J. Lewis, and A.N. Houghton, J. Immunol., 170, 5188 (2003)
Catalog No. 50-168-6396
missing translation for 'QuantityLeft'