Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L 012 sodium salt, Tocris Bioscience™
GSA_VA

Catalog No. 508510 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemiluminescent ROS and RNS indicator

Luminal-based chemiluminescent probe. Detects NAPDH oxidase-derived reactive oxygen and nitrogen species (ROS and RNS). Active in vitro and in vivo.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 143556-24-5
Molecular Formula C13H9ClN4NaO2
Molecular Weight (g/mol) 311.681
InChI Key WRVLDBOUZVBBCX-UHFFFAOYSA-N
Synonym l 012 sodium salt
PubChem CID 131848063
IUPAC Name 8-amino-5-chloro-7-phenyl-2,3-dihydropyrido[3,4-d]pyridazine-1,4-dione;sodium
SMILES C1=CC=C(C=C1)C2=C(C3=C(C(=O)NNC3=O)C(=N2)Cl)N.[Na]

Specifications

Quantity 10 mg
Formula Weight 310.67
Chemical Name or Material L 012 sodium salt
Recommended Storage Store at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.