Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
L-365,260, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
Selective cholecystokinin receptor 2 (CCK2) antagonist (IC50 values are 2 and 280 nM at CCK2 and CCK1 receptors respectively) that is inactive at a range of other receptors including opiate, muscarinic acetylcholine, α- and β adrenergic, histamine, angiotensin and bradykinin receptors.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 398.46 |
Percent Purity | >99% |
Chemical Name or Material | L-365, 260 |
Safety and Handling
Recommended Storage | Store at +4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction