Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-690,330, Tocris Bioscience™

Catalog No. p-4783567 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Inositol monophosphatase inhibitor

A potent inhibitor of inositol monophophatase; stable to hydrolysis. Induces autophagy in COS-7 cells independently of mTOR inhibition.

Chemical Identifiers

CAS 142523-38-4
Molecular Formula C8H12O8P2
Molecular Weight (g/mol) 298.124
InChI Key JKOCAAWWDVHWKB-UHFFFAOYSA-N
Synonym chembl34819, 1-4-hydroxyphenoxy-1-phosphonoethyl phosphonic acid, 1-4-hydroxyphenoxy-1-phosphonoethylphosphonic acid, 1-p-hydroxyphenoxy ethylenebisphosphonate, d06jat, 1-4-hydroxyphenoxy ethylenebisphosphonic acid, 1-4-hydroxyphenoxy ethylidene bisphosphonic acid, phosphonic acid, 1-4-hydroxyphenoxy ethylidene bis, 1-4-hydroxy-phenoxy-1-phosphono-ethyl-phosphonic acid, 1-4-hydroxyphenoxy-1,1-ethanediyl bis phosphonic acid
PubChem CID 132449
IUPAC Name [1-(4-hydroxyphenoxy)-1-phosphonoethyl]phosphonic acid
SMILES CC(OC1=CC=C(C=C1)O)(P(=O)(O)O)P(=O)(O)O

Specifications

Quantity 10 mg
Formula Weight 298.13
Percent Purity >98%
Chemical Name or Material L-690, 330
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.