Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
L-741,626, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
A potent D2 dopamine receptor selective antagonist, with affinities of 2.4, 100 and 220 nM for D2, D3 and D4 receptors respectively. Centrally active following systemic administration in vivo.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 340.85 |
Percent Purity | >98% |
Chemical Name or Material | L-741, 626 |
Safety and Handling
Recommended Storage | Store at Room Temperature |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction