Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-741,742 hydrochloride, Tocris Bioscience™
GSA_VA

Catalog No. p-4783576 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Highly selective D4 antagonist

A potent and highly selective D4 dopamine receptor antagonist, with Ki values of > 1700, 770 and 3.5 nM at cloned human D2, D3 and D4 receptors.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 874882-93-6
Molecular Formula C23H26Cl2N2O
Molecular Weight (g/mol) 417.374
InChI Key HZRPUQURUAXOHB-UHFFFAOYSA-N
Synonym l-741,742 hydrochloride, 5-4-chlorophenyl-4-methyl-3-1-2-phenylethyl piperidin-4-yl isoxazole hydrochloride, 4-5-4-chlorophenyl-4-methylisoxazol-3-yl-1-2-phenylethyl piperidine hydrochloride, 5-4-chlorophenyl-4-methyl-3-1-phenethylpiperidin-4-yl isoxazole hydrochloride, 4-5-4-chlorophenyl-4-methyl-1,2-oxazol-3-yl-1-2-phenylethyl piperidine hydrochloride
PubChem CID 24744861
IUPAC Name 5-(4-chlorophenyl)-4-methyl-3-[1-(2-phenylethyl)piperidin-4-yl]-1,2-oxazole;hydrochloride
SMILES CC1=C(ON=C1C2CCN(CC2)CCC3=CC=CC=C3)C4=CC=C(C=C4)Cl.Cl

Specifications

Quantity 10 mg
Formula Weight 417.38
Percent Purity >98%
Chemical Name or Material L-741, 742 hydrochloride
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.