Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
L-745,870 trihydrochloride, Tocris Bioscience™
Shop All R&D Systems Products

Click to view available options
Quantity:
10 mg
50 mg
Description
A highly potent and selective D4 dopamine receptor antagonist; L-745,870 has Ki values of 0.51, 2300 and 960 nM for D4, D3 and D2 subtypes respectively and > 1000-fold selectivity over 5-HT2, D1 and D5 receptors.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 436.21 |
Percent Purity | >99% |
Chemical Name or Material | L-745, 870 trihydrochloride |
Safety and Handling
Recommended Storage | Desiccate at +4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction