Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
L-748,337, Tocris Bioscience™
Shop All R&D Systems Products

Click to view available options
Quantity:
10 mg
50 mg
Description
Competitive β3-adrenoceptor antagonist that displays selectivity over β1 and β2 receptors (Ki values are 4.0, 204 and 390 nM for β3-, β2- and β1-adrenoceptors respectively). Inhibits cAMP accumulation in response to isoproterenol (IC50 = 6 nM). Reduces iNOS expression, attenuates nitric oxide-induced cell proliferation and induces apoptosis in a melanoma cell line.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 497.61 |
Percent Purity | >98% |
Chemical Name or Material | L-748, 337 |
Safety and Handling
Recommended Storage | Store at +4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction