Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
L-838,417, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
Subtype-selective GABAA receptor partial agonist. Selectively binds to α1, α2, α3 and α5 subunits (Ki values are 0.79, 0.67, 0.67 and 2.25 nM respectively) but displays no efficacy at α1 (α1-sparing). Exhibits non-sedative anxiolytic, antinociceptive and anti-inflammatory activity in vivo.
Specifications
Specifications
Quantity | 10 mg |
Synonym | unii-8czo0970g3, 3-2,5-difluorophenyl-7-1,1-dimethylethyl-6-1-methyl-1h-1,2,4-triazol-5-yl methoxy-1,2,4-triazolo 4,3-b pyridazine, 7-tert-butyl-3-2,5-difluorophenyl-6-1-methyl-1h-1,2,4-triazol-5-yl methoxy-1,2,4 triazolo 4,3-b pyridazine, 5-7-tert-butyl-3-2,5-difluorophenyl-1,2,4 triazolo 4,3-b pyridazin-6-yl oxy methyl-1-methyl-1,2,4-triazole, 1,2,4-triazolo 4,3-b pyridazine, 3-2,5-difluorophenyl-7-1,1-dimethylethyl-6-1-methyl-1h-1,2,4-triazol-5-yl methoxy, l hplc, 1,2,4-triazolo 4,3-b pyridazine,3-2,5-difluorophenyl-7-1,1-dimethylethyl-6-1-methyl-1h-1,2,4-triazol-5-yl methoxy, 3-2,5-difluorophenyl-7-1,1-dimethylethyl-6-2-methyl-2h-1,2,4-triazol-3-ylmethoxy-1,2,4-triazolo 4,3-b pyridazine, 3-2,5-difluorophenyl-7-1,1-dimethylethyl-6-1-methyl-1h-1,2,4,-triazol-5-yl methoxy-1,2,4-triazolo 4,3-b pyridazine |
Formula Weight | 399.4 |
Percent Purity | >98% |
Chemical Name or Material | L-838, 417 |
Safety and Handling
Recommended Storage | Store at +4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction