Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Ascorbic Acid 99.0+%, TCI America™
SDP

Catalog No. A0537500G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:

Chemical Identifiers

CAS

50-81-7

Molecular Formula

C6H8O6

Molecular Weight (g/mol)

176.12

MDL Number

MFCD00064328

InChI Key

CIWBSHSKHKDKBQ-DOMZIZNONA-N

Synonym

l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin

PubChem CID

54670067

ChEBI

CHEBI:29073

IUPAC Name

(5R)-5-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2,5-dihydrofuran-2-one

SMILES

OC[C@H](O)[C@H]1OC(=O)C(O)=C1O

Specifications

Melting Point

192°C

Color

White

Quantity

500 g

Formula Weight

176.12

Percent Purity

≥99.0% (T)

Physical Form

Crystalline Powder

Chemical Name or Material

L-Ascorbic Acid

EINECSNumber

(5)-0062

RTECSNumber

CI7650000

TSCA

Yes

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.