Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ L.b. Cas12a Nuclease
SDP

Catalog No. p-200087432
Click to view available options
Quantity:
200 pmol
2000 pmol

L.b. Cas12a Nuclease for simple, quick and specific analysis of your samples.

L. b. (Lachnospiraceae bacterium) Cas12a Nuclease, also known as Cpf1, is an RNA-guided endonuclease derived from the Class II type V CRISPR-Cas system. It exhibits a high degree of specificity in targeting and cleaving particular DNA sequences in genome editing. Additionally, Cas12a nuclease possesses collateral cleavage activity, resulting in the non-specific cleavage of neighboring DNA upon target recognition.

The DNA cleavage capability of Cas12a nuclease has been leveraged for various applications, including genome editing, gene regulation, and nucleic acid detection. By programming Cas12a with specific guide RNAs, targeted modifications such as gene knockout, gene insertion, or point mutations can be introduced into the target DNA sequence.

Specifications

Product Type Stand-alone enzyme
Form Liquid
Quantity 200 pmol
Research Category Nucleic Acid Analysis
Concentration 10 μM

For Research Use Only.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.