Learn More
Bioworld Lycopersicon esculentum (Tomato) Lectin (LEL/LEA) - Rhodamine (TRITC)
Supplier: Bioworld 217610311
Lycopersicon esculentum (LEA) lectin purified from tomato fruit conjugated to Rhodamine (TRITC).Technical Specifications:Ligand Source:Lycopersicon esculentumMolecular Weight:71 kDaCarbohydrate Specificity:(GlcNAcβ4)n , (Galβ4GlcNAc)nInhibiting/Eluting Sugar:Chitin hydrolysateCarbohydrate for Elution:0.2MChitin hydrolysateBlood Group Specificity:N/AConjugate: RhodamineForm:LiquidIsoelectric (pI):>9Storage: -20°CApplication note: N/A
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.