Learn More
Gold Biotechnology Inc L-Glutathione, Reduced 25 g
SDP
Supplier:  Gold Biotechnology Inc G15525
Encompass_Preferred
L-Glutathione is a co-substrate to glutathione transferase found in plants and animals and is involved in the elimination of free radicals resulting from both cellular respiratory and metabolic activities.This antioxidant plays a role in reducing inflammatory stress in plasma and lymphocytes by directly reacting with reactive oxygen species (ROS). It also serves in amino acid transport and the maintenance of protein sulfhydryl reduction.
Catalog No. NC2943348
missing translation for 'QuantityLeft'
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.