Learn More
Sigma Aldrich Fine Chemicals Biosciences L-Histidine, 71-00-1, MFCD00064315, 100 g
Supplier:  Sigma Aldrich Fine Chemicals Biosciences H6034100G
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass
Empirical Formula (Hill Notation): C6H9N3O2, Molecular Weight: 155.15, cell culture tested, meets EP, USP testing specifications, from non-animal source, Synonym: (S)-2-Amino-3-(4-imidazolyl)propionic acid, NSC 137773. L-histidine has been used as a component in agar plates containing 5-fluorocytosine (5-FC) for the selection of yeast transformants. It has also been used as a component of adult mouse keratinocyte growth medium.
Catalog No. NC1235499
missing translation for 'QuantityLeft'