Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Lactic Acid (Free Acid), MP Biomedicals™

Catalog No. p-4679875 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
1 g
25 g

Chemical Identifiers

CAS 79-33-4
Molecular Formula C3H6O3
Molecular Weight (g/mol) 90.078
MDL Number MFCD00064266
InChI Key JVTAAEKCZFNVCJ-REOHCLBHSA-N
Synonym l-lactic acid, l-+-lactic acid, s-2-hydroxypropanoic acid, s-lactic acid, sarcolactic acid, s-2-hydroxypropionic acid, 2s-2-hydroxypropanoic acid, +-lactic acid, espiritin, tisulac
PubChem CID 107689
ChEBI CHEBI:422
IUPAC Name (2S)-2-hydroxypropanoic acid
SMILES CC(C(=O)O)O

Specifications

Color White
Formula Weight 90.08
Density 1190 to 1250kg/mL
Quantity 1 g
Physical Form Solid
Chemical Name or Material L-Lactic Acid, Free Acid
Recommended Storage Store at 0°C.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.