Learn More
Thomas Scientific L-Shaped Cell Spreader, PP, Individually Wrapped, Sterile, 100/cs
Supplier: Thomas Scientific 1207N33
L-Shaped Cell Spreaders are ideal for evenly spreading bacterial cultures and are made from high quality polypropylene. They are packaged sterile (gamma irradiated) in individual or bulk, easy-to-open, paper/plastic wrappers.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.