Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-(+)-Tartaric Acid, Granular, Multi-Compendial, N.F., J.T.Baker™

Catalog No. 02004398 Shop All Avantor Products
Click to view available options
Quantity:
Packaging:

Chemical Identifiers

CAS

87-69-4

Molecular Formula

C4H6O6

Molecular Weight (g/mol)

150.086

InChI Key

FEWJPZIEWOKRBE-JCYAYHJZSA-N

Synonym

l-tartaric acid, l-+-tartaric acid, l +-tartaric acid, 2r,3r-2,3-dihydroxysuccinic acid, tartaric acid, 2r,3r-2,3-dihydroxybutanedioic acid, r,r-tartaric acid, +-l-tartaric acid, dextrotartaric acid, l-threaric acid

PubChem CID

444305

ChEBI

CHEBI:15671

IUPAC Name

(2R,3R)-2,3-dihydroxybutanedioic acid

SMILES

C(C(C(=O)O)O)(C(=O)O)O

Specifications

Formula Weight

150.09

Physical Form

Granular

Grade

Multi-Compendial/NF

Packaging

Poly Bottle

Specific Gravity

1L = 1.76kg

Quantity

2.5kg

Chemical Name or Material

L-(+)-Tartaric Acid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.