Learn More
Cytiva Laboratory Manifold Replacement Valve (1/pkg)
Supplier:  Cytiva 4894
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
A unique microbiology manifold that allows you to optimize testing without sacrificing cleanliness. With interchangeable components and coupling devices, the manifold takes direction from you.-Microbial cross contamination of your sample is a thing of the past. The manifold is designed to fit most laboratory autoclaves by separating into manageable components.-A larger manifold is at your reach with just a coupler. Modular design allows two manifolds to combine yielding a standard 6-place manifold which can be easily separated for disinfection and/or sterilization.-Whether you are right or left-handed, setting up your test workflow is second nature. The components interchangeable and designed with a simple slip fit to the manifold.- Manifold includes 3-place manifold base, 3 manifold valves, 1 end cap, and 1 hose barb cap. Adapters are required and sold separately This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3116
missing translation for 'QuantityLeft'