Learn More
Research Products International Corp Lab Roller Tube Rotator, with 36 x 1.5 ml Rack
SDP
Supplier:  Research Products International Corp 248140
Encompass
Designed for mixing a variety of popular size tubes from 1.5 ml up to 50 ml. Interchangeable racks allow quick and convenient loading of different size tubes. Racks can be installed to rotate in either horizontal or vertical position at a fixed speed of 28 rpm. Can be operated at temperatures from +4?C to +65?C. Supplied with one rack that holds 36 x 1.5 ml tubes.
Catalog No. 50-550-289
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.